Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K6, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-109 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-109 Supplier Abnova Corporation Supplier No. H00009064A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MAP3K6.

This gene encodes a member of the serine/threonine protein kinase family. The encoded kinase was identified by its interaction with MAP3K5/ASK, a protein kinase and an activator of c-Jun kinase (MAPK7/JNK) and MAPK14/p38 kinase. This kinase was found to weakly activate MAPK7, but not MAPK1/ERK or MAPK14. An alternatively spliced transcript variant has been found for this gene, but its biological validity has not been determined. [provided by RefSeq

Sequence: HFWLHFLLQSCQPFKTACAQGDQCLVLVLEMNKVLLPAKLEVRGTDPVSTVTLSLLEPETQDIPSSWTFPVASICGVSASKRDERCCFLYALPPAQDVQLC

Specifications

Antigen MAP3K6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MAP3K6.
Formulation 50% glycerol
Gene MAP3K6
Gene Accession No. BC015914
Gene Alias ASK2/MAPKKK6/MGC125653/MGC20114
Gene Symbols MAP3K6
Host Species Mouse
Immunogen MAP3K6 (AAH15914, 225 a.a. ∼ 325 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9064
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.