Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK 1/2 (tERK) Rabbit anti-Human, AlexaFluor™647, Polyclonal, MilliporeSigma™

Catalog No. FCABS165A6 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCABS165A6 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCABS165A6 Supplier MilliporeSigma Supplier No. FCABS165A6
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Milli-Mark™ Anti-MAPK 1/2 (tERK) Antibody, Alexa Fluor 647 Conjugate detects level of MAPK 1/2 (tERK) and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPK 1/2 (tERK)
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation Purified rabbit polyclonal antibody conjugated to Alexa Fluor 647 in PBS buffer with 15mg/mL BSA and 0.1% Sodium Azide
Gene Symbols MAPK1; p38; ERK; P42MAPK; ERK2; p40; PRKM1; p41mapk; PRKM2; MAPK2; p42-MAPK; p41; ERK-2; ERT1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to rat MAPK 1/2 at the C-terminus.
Purification Method Protein A purfied
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) NP_059043
Target Species Human
Content And Storage 2-8°C. Protected from light in undiluted aliquots for 6 months.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.