Learn More
mitogen-activated protein kinase 14, Mouse, Clone: 3D5, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is activated by various environmental stresses and proinflammatory cytokines. The activation requires its phosphorylation by MAP kinase kinases (MKKs), or its autophosphorylation triggered by the interaction of MAP3K7IP1/TAB1 protein with this kinase. The substrates of this kinase include transcription regulator ATF2, MEF2C, and MAX, cell cycle regulator CDC25B, and tumor suppressor p53, which suggest the roles of this kinase in stress related transcription and cell cycle regulation, as well as in genotoxic stress response. Four alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
Specifications
| Antigen | mitogen-activated protein kinase 14 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, In situ PLA, Western Blot |
| Classification | Monoclonal |
| Clone | 3D5 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant MAPK14. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MAPK14 |
| Gene Accession No. | BC031574 |
| Gene Alias | CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.