Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mitogen-activated protein kinase 6, Mouse, Clone: 4C11, Abnova™

Catalog No. 89000900 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-900 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-900 Supplier Abnova Corporation Supplier No. H00005597M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MAPK6.

The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters. [provided by RefSeq

Sequence: EVRKDEQVEKENTYTSYLDKFFSRKEDTEMLETEPVEDGKLGERGHEEGFLNNSGEFLFNKQLESIGIPQFHSPVGSPLKSIQATLTPSAMKSSPQIPHQTYSSILKHLN

Specifications

Antigen mitogen-activated protein kinase 6
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MAPK6.
Formulation PBS with no preservative; pH 7.4
Gene MAPK6
Gene Accession No. BC035492
Gene Alias DKFZp686F03189/ERK3/HsT17250/PRKM6/p97MAPK
Gene Symbols MAPK6
Host Species Mouse
Immunogen MAPK6 (AAH35492, 612 a.a. ∼ 721 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5597
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.