Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAPK-2 Mouse, PE, Clone: P24-694, BD

Catalog No. BDB562470 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562470 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562470 Supplier BD Biosciences Supplier No. 562470
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The P24-694 monoclonal antibody specifically binds to the phosphorylated T334 site (pT334) of MAPKAPK-2. MAPKAPK-2 is a serine/threonine protein kinase. This ∽49 kDa member of the MAPKAPK family of protein kinases is also known as mitogen-activated protein kinase-activated protein kinase 2. MAPKAPK-2 is phosphorylated and activated by p38 MAP kinase in response to stress, cytokines and chemokines. MAPKAPK-2 is phosphorylated on multiple sites including Thr222, Ser272 and Thr334. Phosphorylation of any two of these three amino acid residues seems to be required for the activation of this kinase thatserves multiple cellular functions. Phosphorylation of Thr334 was reported to be essential for nuclear export of the heterodimer formed between p38 MAPK and MAPKAPK-2. Mice deficient in MAPKAPK-2 have been shown to be protected from ischemic injury. MAPKAPK-2 is also reported to serve as a cell cycle checkpoint kinase in response to UV irradiation. The heat shock protein, HSP27 was shown to be one of the major substrates of MAPK and MAPKAPK-2.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPKAPK-2
Applications Flow Cytometry
Classification Monoclonal
Clone P24-694
Conjugate PE
Description MAP kinase-activated protein kinase 2, MAPKAP kinase 2; MK2; MAPK2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated human MAPKAPK-2 peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.