Learn More
microtubule-associated protein, RP/EB family, member 2 (B02), Mouse anti-Human, Rat, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MAPRE2 protein.
Supplier: Abnova Corporation H00010982B02
Description
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: MPGPTQTLSPNGENNNDIIQDNNGTIIPFRKHTVRGERSYSWGMAVNVYSTSITQETMSRHDIIAWVNDIVSLNYTKVEQLCSGAAYCQFMDMLFPGCISLKKVKFQAKLEHEYIHNFKLLQASFKRMNVDKVIPVEKLVKGRFQDNLDFIQWFKKFYDANYDGKEYDPVEARQGQDAIPPPDPGEQIFNLPKKSHHANSPTAGAAKSSPAAKPGSTPSRPSSAKRASSSGSASKSDKDLETQVIQLNEQVHSLKLALEGVEKERDFYFGKLREIELLCQEHGQENDDLVQRLMDILYASEEHEGHTEEPEAEEQAHEQQPPQQEEYSpecifications
microtubule-associated protein, RP/EB family, member 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MAPRE2 protein. | |
MAPRE2 | |
EB1/EB2/RP1 | |
Mouse | |
50 μL | |
Yes | |
10982 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_014268.1 | |
MAPRE2 | |
MAPRE2 (NP_055083.1, 1 a.a. ∼ 327 a.a) full-length human protein. | |
RUO | |
Primary | |
Human, Rat | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.