Learn More
MARCH8, clone: 710424, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524329
Description
MARCH8 (c-MIR) is a novel E3 ubiquitin ligase designated as the modulator of immune recognition (MIR) family, whose catalytic domain is a variant RING domain (RING-CH domain). MARCH8 was found as a functional and structural homolog of KSHV MIR1 and MIR2. MARCH8 targets B7-2 to lysosomal degradation and down-regulates the B7-2 surface expression through ubiquitination of its cytoplasmic tail. Furthermore, MARCH8 has been shown to down-regulate the expression of transferrin receptor and Fas, an important molecule for the induction of apoptosis. MARCH8 is the first example of an E3 ubiquitin ligase that is capable of inhibiting MHC II expression. Recent findings suggest that MARCH8 may regulate immune responses by promoting ubiquitination of MHC-II and CD86, leading to their subsequent endocytosis and lysosomal degradation.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
MARCH8 | |
Monoclonal | |
0.5mg/mL | |
PBS with 5% trehalose and no preservative | |
Q5T0T0, Q9DBD2 | |
MARCH8 | |
E. coli-derived recombinant human MARCH8 Gln218-Val291 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Western Blot | |
710424 | |
Unconjugated | |
MARCH8 | |
RP11-67C2.1, MARCH-VIII, MIR, RNF178, c-MIR, E3 ubiquitin-protein ligase MARCH8, RING finger protein 178, c-mir, cellular modulator of immune recognition, cellular modulator of immune recognition (c-MIR), membrane-associated RING finger protein 8, membrane-associated RING-CH protein VIII | |
Mouse | |
Protein A/G | |
RUO | |
220972, 312656, 71779 | |
Store at-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.