Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK2, Mouse, Clone: 3C5, Abnova™

Catalog No. 89021441 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-441 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-441 Supplier Abnova Corporation Supplier No. H00002011M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MARK2.

This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: QAAGPAIPTSNSYSKKTQSNNAENKRPEEDRESGRKASSTAKVPASPLPGLERKKTTPTPSTNSVLSTSTNRSRNSPLLERASLGQASIQNGKDSLTMPG

Specifications

Antigen MARK2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MARK2.
Formulation PBS with no preservative; pH 7.4
Gene MARK2
Gene Accession No. BC008771
Gene Alias EMK1/MGC99619/PAR-1/Par1b
Gene Symbols MARK2
Host Species Mouse
Immunogen MARK2 (AAH08771, 401 a.a. ∼ 500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2011
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.