Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MASH1 Mouse, Unlabeled, Clone: 24B7.2D11, BD

Catalog No. BDB556604 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556604 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556604 Supplier BD Biosciences Supplier No. 556604
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MASH1 and MASH2 (Mammalian achaete-schute Homolog 1 and 2) are basic helixloop-helix transcription factors which are mammalian homologs of the achaete-schute gene family that is required for neuronal development in Drosophila. The bHLH motif is present in many types of transcription factors including E2-2, E47, MyoD and others. MASH1 has been shown to heterodimerize with other bHLH containing transcription factors. MASH1-E12 heteromers bind to and promote transcriptional activation of muscle creatine kinase, a known target for MyoD activation. However, MASH1 appears to play its primary role during early development of the autonomic nervous system. In committed neuronal precursor cells, early expression of MASH1 activates a subset of neuron-specific genes to promote differentiation. Thus null mutations in the MASH1 gene suggest that MASH1 is a valuable marker for investigation of neural development in mammals. MASH1 is observed as an ∽34kDa protein by SDS-PAGE.

Host Species: Mouse
Clone: 24B72D11.1
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat MASH1 full length recombinant protein

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MASH1
Applications Western Blot
Classification Monoclonal
Clone 24B7.2D11
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat MASH1 full length recombinant protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.