Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maspin Mouse anti-Human, Unlabeled, Clone: G167-70, BD

Catalog No. BDB554292 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554292 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554292 Supplier BD Biosciences Supplier No. 554292
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Maspin is a 42kDa protein that may play a role in human breast cancer by functioning as a tumor suppressor. Maspin is expressed in normal mammary epithelial cells, but there is a loss of expression in invasive breast cancer including lymph node and distant metastases. Further, maspin is not expressed in most mammary carcinoma cell lines. Mammary carcinoma cells transfected with recombinant maspin have a reduced ability to migrate through reconstituted basement membrane and lose their ability to form tumors in nude mice, providing support for maspin as a tumor suppressor protein. Maspin has homology to members of the serpin family of protease inhibitors, some of which have been implicated in the regulation of invasion and metastasis. Yet protease inhibitory activity has not been detected using recombinant maspin. It has been suggested that maspin, unlike most serpins, may be a ligand-binding serpin rather than a protease inhibitor. However, the question of whether or not maspin has protease inhibitor activity remains to be determined. G167-70 is specific for human maspin. It detects maspin as a 42kDa protein in western blots of normal human mammary epithelial cells. Many mammary tumor cell lines including MDA-MB-231 (ATCC
HTB 26) and MDA-MB-435S (ATCC HTB 129) lack maspin expression and are suitable as negative controls. Purified recombinant human maspin from a yeast expression system was used as immunogen.

Host Species: Mouse
Clone: G167-70
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Recombinant Human Maspin
Formula Weight [Chemical]: 42kDa

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Maspin
Applications Western Blot
Classification Monoclonal
Clone G167-70
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human Maspin
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.