Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAST1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528583
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.
Specifications
MAST1 | |
Polyclonal | |
Unconjugated | |
MAST1 | |
9430008B02Rik; KIAA0973; MAST1; mast1 {ECO:0000312; microtubule associated serine/threonine kinase 1; microtubule-associated serine/threonine-protein kinase 1; RGD:631372}; SAST; SAST170; syntrophin associated serine/threonine kinase; syntrophin-associated serine/threonine-protein kinase; syntrophin-associated serine-threonine protein kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22983, 56527 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9R1L5, Q9Y2H9 | |
MAST1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human MAST1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction