Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATK Mouse anti-Human, Clone: 9D7, MilliporeSigma™

Catalog No. MABC682MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC682MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC682MI Supplier MilliporeSigma Supplier No. MABC682
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Megakaryocyte-associated tyrosine-protein kinase using this mouse monoclonal antibody, Anti-MATK Antibody, clone 9D7 validated for use in western blotting and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATK
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 9D7
Conjugate Unconjugated
Formulation Purified mouse monoclonal liquid in PBS containing up to 0.1% sodium azide and 50% glycerol.
Gene Symbols MATK, CTK, HYL
Host Species Mouse
Immunogen Purified recombinant fragment of human MATK expressed in E. Coli.
Purification Method Antigen Affinity purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.