Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MB67 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA536268
Description
This antibody detects endogenous protein at a molecular weight of 35 kDa. Purity is >95% by SDS-PAGE.
This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists. Ligand binding results in translocation of this protein to the nucleus, where it activates or represses target gene transcription. These ligands include bilirubin, a variety of foreign compounds, steroid hormones, and prescription drugs. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
MB67 | |
Polyclonal | |
Unconjugated | |
NR1I3 | |
Nuclear receptor subfamily 1 group I member 3, Constitutive activator of retinoid response, Constitutive active response, Constitutive androstane receptor, CAR, Orphan nuclear receptor MB67, NR1I3, CAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12355, 65035, 9970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14994, O35627, Q9QUS1 | |
NR1I3 | |
Synthetic peptide corresponding to amino acids 70-124 of human MB67 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction