Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

methyl-CpG binding domain protein 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005735 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-735 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-735 Supplier Abnova Corporation Supplier No. H00004152B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MBD1 protein.

DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. Five transcript variants of the MBD1 are generated by alternative splicing resulting in protein isoforms that contain one MBD domain, two to three cysteine-rich (CXXC) domains, and some differences in the COOH terminus. All five transcript variants repress transcription from methylated promoters; in addition, variants with three CXXC domains also repress unmethylated promoter activity. MBD1 and MBD2 map very close to each other on chromosome 18q21. [provided by RefSeq

Sequence: MAEDWLDCPALGPGWKRREVFRKSGATCGRSDTYYQSPTGDRIRSKVELTRYLGPACDLTLFDFKQGILCYPAPKAHPVAVASKKRKKPSRPAKTRKRQVGPQSGEVRKEAPRDETKADTDTAPASFPAPGCCENCGISFSGDGTQRQRLKTLCKDCRAQRIAFNREQRMFKRVGCGECAACQVTEDCGACSTCLLQLPHDVASGLFCKCERRRCLRIVERSRGCGVCRGCQTQEDCGHCPICLRPPRPGLRRQWKCVQRRCLRGKHARRKGGCDSKMAARRRPGAQPLPPPPPSQSPEPTEPHPRALAPSPPAEFIYYCVDEDELQPYTNRRQNRKCGACAACLRRMDCGRCDFCCDKPKFGGSNQKRQKCRWRQCLQFAMKRLLPSVWSESEDGAGSPPPYRRRKRPSSARRHHLGPTLKPTLATRTAQPDHTQAPTKQEAGGGFVLPPPGTDLVFLREGASSPVQVPGPVAASTEALLQEAQCSGLSWVVALPQVKQEKADTQDEWTPGTAVLTSPVLVPGCPSKAVDPGLPSVKQEPPDPEEDKEENKDDSASKLAPEEEAGGAGTPVITEIFSLGGTRFRDTAVWLPRSKDLKKPGARKQ

Specifications

Antigen methyl-CpG binding domain protein 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MBD1 protein.
Formulation No additive
Gene MBD1
Gene Accession No. NM_015846.2
Gene Alias CXXC3/PCM1/RFT
Gene Symbols MBD1
Host Species Mouse
Immunogen MBD1 (NP_056671.2, 1 a.a. ∼ 605 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.