Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 072054MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-205-4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-205-4MI Supplier MilliporeSigma Supplier No. 072054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-MBD1 Antibody, C-terminus is a Rabbit Polyclonal Antibody for detection of MBD1 also known as CXXC-type zinc finger protein 3 and has been validated in western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.09% NaN3.
Gene Symbols CXXC3; OTTHUMP00000163504; OTTHUMP00000163506; OTTHUMP00000163507; PCM1 ; RFT
Host Species Rabbit
Immunogen The immunogen was a KLH-conjugated synthetic peptide corresponding the C-terminal region of human MBD1.
Purification Method Protein G chromatography
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_079103
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.