Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBNL2 Mouse anti-Mouse, Clone: MB2a(3B4), MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE959
Description
This Anti-MBNL2 Antibody, clone MB2a(3B4) is validated for use in Western Blotting for the detection of MBNL2.
Specifications
MBNL2 | |
Monoclonal | |
1mg/mL | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
MBNL2, MBLL, MBLL39, MLP1 | |
Recombinant protein corresponding to human MBNL2. | |
100 μg | |
Mouse | |
Purified |
Western Blot | |
MB2a(3B4) | |
Unconjugated | |
NP_659002 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction