Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Mouse anti-Human, Mouse, Rat, Clone: SKB3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05675
Description
Detect MBP using this Anti-MBP Antibody, clone SKB3 validated for use in western blotting.
Specifications
MBP | |
Monoclonal | |
Unconjugated | |
NM_002385 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Western Blot | |
SKB3 | |
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.02M Phosphate Buffer, 0.25MNaCl, pH 7.6 with 15mg/mL BSA containing no preservatives. | |
MBP, MGC99675 | |
Peptide corresponding to the human myelin basic protein sequence containing amino acid Thr98 | |
50 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction