Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM Mouse anti-Canine, Human, Mouse, Biotin, Clone: P1H12, MilliporeSigma™

Catalog No. MAB16985BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB16985BMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB16985BMI Supplier MilliporeSigma Supplier No. MAB16985B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MCAM Antibody, clone P1H12, biotin conjugated is validated for use in ELISA, flow cytometry, immunoprecipitation, immunocytochemistry, immunohistochemistry for the detection of MCAM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAM
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone P1H12
Conjugate Biotin
Formulation Purified immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl, pH=7.6, 15 mg/nL BSA containing 0.1% sodium azide as a preservative.
Gene Accession No. NM_006500.2
Gene Symbols MCAM, Gicerin, MUC18, CD146
Host Species Mouse
Immunogen Immunization of mice with HUVEC
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.