Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM Mouse anti-Chicken, Turkey, Unconjugated, Clone: C264, Southern Biotech™
SDP

Catalog No. OB838501 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB838501 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB838501 Supplier Southern Biotechnology Associates Supplier No. 838501
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.

Specifications

Antigen MCAM
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone C264
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
Host Species Mouse
Immunogen Mixture of E15 and E16 thymocytes from H.B15, H.B19, and H.B21 chicken embryos
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken, Turkey
Content And Storage Store at 2-8°C
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.