Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM Mouse anti-Canine, Human, Mouse, Clone: P1H12, MilliporeSigma™

Catalog No. MAB16985MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB16985MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB16985MI Supplier MilliporeSigma Supplier No. MAB16985
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MCAM Antibody, clone P1H12 detects level of MCAM and has been published and validated for use in ELISA, flow cytometry, immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAM
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone P1H12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.02MPB with 0.25MNaCl, pH=7.6, with 0.1% sodium azide.
Gene Accession No. NM_006500.2
Gene Symbols MCAM, Gicerin, MUC18, CD146
Host Species Mouse
Immunogen Immunization of mice with HUVEC.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.