Learn More
MilliporeSigma™ anti-MCAM (MUC18; CD146), Clone: P1H12, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB16985B/DEL
Description
MUC18 antigen (CD146), a member of the immuoglobulin superfamily, is an intrinsic membrane glycoprotein of 110-120 kDa found on the surface of endothelial cells, bone marrow fibroblasts and various melanomas. MUC18 has been used as a marker of tumor progression in human melanoma because expression in those tumors correlates strongly with poor prognosis and the development of metastic disease. In addition, a number of human T, B and myeloid leukemic cell lines seem to express MUC18. The close structural relationship with N-CAM and related molecules suggests that MUC18 may be also a developmentally regulated cell adhesion molecule (Melanoma adhesion molecule or MCAM).
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
MCAM (MUC18; CD146) | |
Biotin | |
Immunization of mice with HUVEC | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Cell Structure | |
Canine, Human, Mouse, Rabbit | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.