Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCCC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535323
Description
MCCC2 is the small subunit of 3-methylcrotonyl-CoA carboxylase. This enzyme functions as a heterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoA to form 3-methylglutaconyl-CoA.
Specifications
MCCC2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9HCC0, Q3ULD5, Q5XIT9 | |
MCCC2 | |
KLH conjugated synthetic peptide between 163-189 amino acids from the central region of human MCCC2 | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
MCCC2 | |
MCCC2; MCCB; Methylcrotonoyl-CoA carboxylase beta chain, mitochondrial; 3-methylcrotonyl-CoA carboxylase 2; 3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
361884, 64087, 78038 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction