Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCL-1 Mouse anti-Human, Unlabeled, Clone: 22, BD

Catalog No. BDB559027 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559027 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559027 Supplier BD Biosciences Supplier No. 559027
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Bcl-2 family of proteins plays a pivotal role in determining the life and death of a cell. Members of the Bcl-2 family all possess at least one of four conserved Bcl-2 homology domains (BH1-BH4) which mediate protein-protein interactions. They can be divided into molecules that have either an anti-apoptotic or pro-apoptotic function. The ratio between these two groups of molecules can help determine whether a cell lives or dies. The majority of anti-apoptotic family members contain common BH1-BH4 domains and are most similar to Bcl-2. Mcl-1 is a member of the anti-apoptotic Bcl-2 group of proteins, including Bcl-x, Bcl-w, A1/Bfl-1, Boo/Diva, and Nr-13. Mcl-1 was cloned from the ML-1 human myeoblastic leukemia cell line. Mcl-1 localizes to the mitochrondria, like Bcl-2, but also associates with membranes via its C-terminal hydrophobic domain. Further studies have shown that Mcl-1 is upregulated in response to the GM-CSF and IL-3 cytokine signaling pathways and that this regulation of Mcl-1 is controlled at the level of transcription. Mcl-1 is detected at approximately 40kDa in SDS/PAGE.

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCL-1
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified recombinant human Mcl-1 lacking the C-terminal hydrophobic domain
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.