Learn More
Mcl-1 Mouse anti-Human, Clone: RC13, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MABC43
Catalog No. MABC43MI
Only null left
Add to Cart
Description
Description
Detect Mcl-1 using this Anti-Mcl-1 Antibody, clone RC13 validated for use in western blot, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mcl-1
Monoclonal
Unconjugated
NP_068779
Mouse
Protein G Purified
Primary
2-8°C for one year
IgG1 κ
Immunohistochemistry, Immunoprecipitation, Western Blot
RC13
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
MCL1, MCL1L, Mcl-1, mcl1/EAT, TM, MCL1S, EAT
Recombinant protein corresponding to human Mcl-1.
100 μL
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only