Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mcl-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB2910MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2910MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2910MI Supplier MilliporeSigma Supplier No. AB2910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Mcl-1 Antibody is validated for use in immunoprecipitation, western blot, immunohistochemistry (paraffin) for the detection of Mcl-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCl−1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid antiserum.
Gene Symbols MCL1; MCL1L; mcl1/EAT; MGC104264; MGC1839; TM; MCL1S; EAT
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 121-139 of human MCl−1.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_021960.3; NM_182763.1
Target Species Human
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.