Learn More
MCM3, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA121998
Description
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.
PA1-21998 detects MCM3 in Human samples.PA1-21998 has been successfully used in Immunoprecipitation and Western Blot procedures.PA1-21998 immunogen corresponds to Synthetic peptide of 38 amino acids representing a portion of human MCM 3 encoded by exons 10 and 11.
Specifications
MCM3 | |
Polyclonal | |
Unconjugated | |
P25205, P25206 | |
MCM3 | |
Synthetic peptide of 38 amino acids representing a portion of human MCM 3 encoded by exons 10 and 11 | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
1mg/mL | |
MCM3 | |
HCC5, MGC1157, P1-MCM3, P1.h, RLFB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4172 | |
Store at 4°C | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.