Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM6 Mouse anti-Human, Unlabeled, Clone: 1, BD

Catalog No. BDB611622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611622 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611622 Supplier BD Biosciences Supplier No. 611622
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rabbit anti-Goat IgG (H+L) Secondary Antibody, AP conjugate, 61-1622, from Invitrogen™. Species Reactivity: Goat; Applications: ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Host Species: Rabbit
Cross Reactivity: Goat
Isotype: IgG
Immunogen: Gamma Immunoglobin
Formulation: 1mL purified antibody in 50mM tris with 1% BSA, 40% glycerol and 0.1% sodium azide
This product detects Goat IgG (H+L) in Goat samples. This product has been successfully used in ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation and Western Blot procedures. The immunogen is Gamma Immunoglobin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM6
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MCM6 aa. 670-792
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.