Learn More
MCOLN2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human MCOLN2 peptide using ARM Technology.
Supplier: Abnova Corporation H00255231K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Mucolipins constitute a family of cation channel proteins with homology to the transient receptor potential superfamily. In mammals, the mucolipin family includes 3 members, MCOLN1 (MIM 605248), MCOLN2, and MCOLN3 (MIM 607400), that exhibit a common 6-membrane-spanning topology. Homologs of mammalian mucolipins exist in Drosophila and C. elegans. Mutations in the human MCOLN1 gene cause mucolipodosis IV (MIM 262650) (Karacsonyi et al., 2007 [PubMed 17662026]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| MCOLN2 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| MCOLN2 | |
| MCOLN2 | |
| A synthetic peptide of human MCOLN2 | |
| RUO | |
| 255231 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| mucolipin 2 | |
| MCOLN2 | |
| FLJ36691, TRP-ML2, TRPML2 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.