Learn More
MCP-1 Mouse anti-Human, PE, Clone: 5D3-F7, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559324
Description
The 5D3-F7 antibody reacts with human monocyte chemoattractant protein-1 (MCP-1), also known as monocyte chemotactic and activating factor (MCAF). This antibody is routinely tested by ELISA Detection. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Intracellular Staining

Specifications
MCP-1 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Recombinant Human MCP-1 | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
5D3-F7 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.