Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCP-1 Mouse anti-Human, Biotin, Clone: 5D3-F7, BD

Catalog No. BDB554664 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554664 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554664 Supplier BD Biosciences Supplier No. 554664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5D3-F7 antibody reacts with human monocyte chemoattractant protein-1 (MCP-1), also known as monocyte chemotactic and activating factor (MCAF). This antibody is routinely tested by ELISA Detection. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 5D3-F7
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant Human MCP-1

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Applications ELISA
Classification Monoclonal
Clone 5D3-F7
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human MCP-1
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.