Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCP-1 Mouse anti-Human, Unlabeled, Clone: 10F7, BD

Catalog No. BDB555055 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555055 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555055 Supplier BD Biosciences Supplier No. 555055
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 10F7 antibody reacts with human monocyte chemoattractant protein-1 (MCP-1), also known as monocyte chemotactic and activating factor (MCAF). The immunogen used to generate the 10F7 hybridoma was E. coli-expressed recombinant human MCP-1. This antibody is routinely tested by ELISA Capture. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Applications ELISA
Classification Monoclonal
Clone 10F7
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen E-coli-expressed recombinant human MCP-1
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.