Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCP-1 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: 2H5, BD

Catalog No. BDB554440 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554440 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554440 Supplier BD Biosciences Supplier No. 554440
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 2H5 antibody reacts with mouse and rat monocyte chemoattractant protein (MCP-1), formerly termed JE. This antibody also recognizes human MCP-1, but shows no reactivity with the closely related mouse β chemokines, TCA3 and MIP-1β. The immunogen used to generate the 2H5 hybridoma was heparin-purified CHO-expressed mouse MCP-1. This is a neutralizing antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Applications ELISA
Classification Monoclonal
Clone 2H5
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Hamster
Immunogen Heparin-purified CHO-expressed mouse MCP-1
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.