Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCP-3 Mouse anti-Human, Unlabeled, Clone: 9H11, BD

Catalog No. BDB551228 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551228 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551228 Supplier BD Biosciences Supplier No. 551228

Mouse Monoclonal Antibody

This antibody reacts with human monocyte chemotactic protein-3 (MCP-3). The immunogen used to produce the 9H11 hybridoma was E. coli-expressed recombinant MCP-3. This is a neutralizing antibody.

Host Species: Mouse
Clone: 9H11
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: E. coli-expressed recombinant MCP-3

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-3
Applications ELISA
Classification Monoclonal
Clone 9H11
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen E. coli-expressed recombinant MCP-3
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.