Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mDia1 Mouse, Unlabeled, Clone: 51, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610849
Description
GTPases like Ras, Rho, cdc42Hs, and Rac modulate a multitude of cellular functions like cytoskeletal architecture, growth, motility, and gene expression. The activity of the GTP-binding proteins is regulated by factors that accelerate GTP-binding (GAPs) and proteins that enhance the rate of GTP hydrolysis. mDia1, also known as p140mDia and Drf1, is the mammalian homologue of Drosophila's diaphanous, a protein essential for cytokinesis. The 1255 amino acid mDia1 is widely expressed, with a Rho binding domain at the NH2-terminal region, a central polyproline region, and an FH2 domain. This protein also shares homology with the yeast Bn1p essential for budding and the mouse formin necessary for proper limb development. mDia1 binds to GTP-Rho and to the actin-binding protein profilin-all three proteins are co-localized at the lamellipodia in cultured cells. The overexpression of mDia1 promoted the formation of actin filaments, implicating this protein in cell motility events regulated by Rho. mDia1 was also found consistently mutated in familial deafness.
Specifications
mDia1 | |
Monoclonal | |
250μg/mL | |
p140mDia; Drf1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
51 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse p140mDia aa. 41-153 | |
150 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction