Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM2 Mouse anti-Human, Clone: 2A10, MilliporeSigma™

Catalog No. MABE281MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE281MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE281MI Supplier MilliporeSigma Supplier No. MABE281
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MDM2 Antibody, clone 2A10 is a Mouse Monoclonal Antibody for detection of MDM2 also known as E3 ubiquitin-protein ligase Mdm2 or Hdm2 and has been validated in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDM2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ cultured supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002383
Gene Symbols MDM2
Host Species Mouse
Immunogen Human MDM2 protein
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.