Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM2 Mouse anti-Human, Mouse, Clone: 4B2C1.11, MilliporeSigma™

Catalog No. MABE331MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE331MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE331MI Supplier MilliporeSigma Supplier No. MABE331
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-MDM2 Antibody, clone 4B2C1.11 (mouse monoclonal antibody) validated in western blot, immunoprecipitation, immunocytochemistry to detect MDM2 also known as E3 ubiquitin-protein ligase Mdm2, Double minute 2 protein, Hdm2, Oncoprotein Mdm2, p53-binding protein Mdm2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDM2
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4B2C1.11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002383
Gene Symbols MDM2
Host Species Mouse
Immunogen Recombinant protein corresponding to human MDM2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.