Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM4 Mouse anti-Human, Mouse, Clone: 7A8, MilliporeSigma™

Catalog No. 041556MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-155-6MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-155-6MI Supplier MilliporeSigma Supplier No. 041556
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-MDM4 Antibody, clone 7A8 (mouse monoclonal antibody) validated in western blotting to detect MDM4 also known as Double minute 4 protein, MDM4-related protein 1, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDM4
Applications Western Blot
Classification Monoclonal
Clone 7A8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_032601
Gene Symbols MDM4, HDMX, MDMX, MRP1
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to mouse MDM4.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.