Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mdmx Mouse anti-Human, Clone: 8C6, MilliporeSigma™

Catalog No. 041555MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-155-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-155-5MI Supplier MilliporeSigma Supplier No. 041555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mdmx Antibody, clone 8C6 is a Mouse Monoclonal Antibody for detection of Mdmx also known as MDM4-related protein 1, Mdm2-like p53-binding protein and has been validated in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mdmx
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8C6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_002384.2
Gene Symbols HDMX, MDMX, MRP1, MDM4
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human Mdmx.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.