Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1 Mouse anti-Human, Clone: 6C4.2, MilliporeSigma™

Catalog No. MAB4338MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4338MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4338MI Supplier MilliporeSigma Supplier No. MAB4338

Mouse Monoclonal Antibody

Detect MDR1 using this Anti-MDR1 Antibody, clone 6C4.2 validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDR1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6C4.2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M PBS pH 7.6, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_000927.3
Gene Symbols ABCB1, PGY1, MGC163296, CD243, GP170, P-gp, MDR1, CLCS, ABC20, EC 3.6.3.44
Host Species Mouse
Immunogen Human MDR-transfected NIH3T3 cells
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.