Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1 Mouse anti-Human, Clone: MC57, MilliporeSigma™

Catalog No. MAB4163MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4163MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4163MI Supplier MilliporeSigma Supplier No. MAB4163
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MDR1 Antibody, extracellular human specific Pgp, clone MC57 detects level of MDR1 and has been published and validated for use in flow cytometry, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDR1
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone MC57
Conjugate Unconjugated
Formulation Purified immunoglobulin from Protein A Sepharose chromatography. Liquid in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NM_000927.3
Gene Symbols ABCB1, PGY1, MGC163296, GP170, ABC20, CD243, P-gp, MDR1, CLCS, EC 3.6.3.44
Host Species Mouse
Immunogen Living intact human multidrug-resistant cells
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.