Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1b Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB10337MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10337MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10337MI Supplier MilliporeSigma Supplier No. AB10337

Rabbit Polyclonal Antibody

Anti-MDR1b Antibody, ATP Binding Cassette Sub family B is an antibody against MDR1b for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDR1b
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Raw rabbit serum with 0.05% sodium azide.
Gene Symbols EC 3.6.3.44; ATP-binding cassette sub-family B member 1; P-glycoprotein 1; CD243 antigen
Host Species Rabbit
Immunogen Recombinant rat MDR1b
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_012623
Target Species Rat
Content And Storage −20°C for 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.