Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR3 Mouse anti-Human, Clone: P3 II-26, MilliporeSigma™

Catalog No. MAB4140MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4140MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4140MI Supplier MilliporeSigma Supplier No. MAB4140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MDR3 Antibody, clone P3 II-26 is an antibody against MDR3 for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDR3
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone P3 II-26
Conjugate Unconjugated
Formulation Liquid containing 0.7% BSA and 0.1% sodium azide.
Gene Accession No. NM_000443.3; NM_018849.2; NM_018850.2
Gene Symbols ABCB4, PGY3, PFIC3, PFIC-3, MDR2/3, MDR3, MDR2, ABC21, GBD1, EC 3.6.3.44
Host Species Mouse
Immunogen Mab was selected after immunization with a fusion protein consisting of Gluthatione-S-Transferase and a fragment of MDR3 P-gp comprising aa 629-692.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months. −20°C for long term.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.