Learn More
malic enzyme 1, NADP(+)-dependent, cytosolic, Mouse, Clone: 3H5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ME1.
Supplier: Abnova Corporation H00004199M03
Description
This gene encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet. [provided by RefSeq
Sequence: MEPEAPRRRHTHQRGYLLTRNPHLNKDLAFTLEERQQLNIHGLLPPSFNSQEIQVLRVVKNFEHLNSDFDRYLLLMDLQDRNEKLFYRVLTSDIEKFMPIVYTPTVGLACQQYSLVFRKPRGLFITIHDRGHIASVLNAWPEDVIKAIVVTDGERILGLGDLGCNGMGIPVGKLALYTACGGMNPQECLPVILDVGTENEELLKDPLYIGLRQRRVRGSEYDDFLDEFMEAVSSKYGMNCLIQFEDFANVNAFRLLNKYRNQYCTFNDDIQGTASVAVAGLLAALRITKNKLSDQTILFQGAGEAALGIAHLIVMALEKEGLPKEKAIKKIWLVDSKGLIVKGRASLTQEKEKFAHEHEEMKNLEAIVQEIKPTALIGVAAIGGAFSEQILKDMAAFNERPIIFALSNPTSKAECSAEQCYKITKGRAIFASGSPFDPVTLPNGQTLYPGQGNNSYVFPGVALGVVACGLRQITDNIFLTTAEVIAQQVSDKHLEEGRLYPPLNTIRDVSLKIAEKIVKDAYQEKTATVYPEPQNKEAFVRSQMYSTDYDQILPDCYSWPEEVQKIQTKVDQSpecifications
malic enzyme 1, NADP(+)-dependent, cytosolic | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC025246 | |
ME1 | |
ME1 (AAH25246, 1 a.a. ∼ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
4199 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
3H5 | |
Mouse monoclonal antibody raised against a full length recombinant ME1. | |
ME1 | |
HUMNDME/MES | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.