Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ME3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539278
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis, metastasis, and atherosclerosis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases.
Specifications
ME3 | |
Polyclonal | |
Unconjugated | |
ME3 | |
Macrophage Metaloelastase, Macrophage Metaloelastase, Matrix Metalloprotease 3, Matrix Metalloprotease 3, MME, MME, MMP3, , HME, Macrophage elastase, ME | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10873, 109264 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q16798, Q8BMF3 | |
ME3 | |
A synthetic peptide derived from the internal region of human ME3 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction