Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Measles Blend Mouse anti-Human, Clone: CV1, CV4, MilliporeSigma™

Catalog No. MAB8905MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8905MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8905MI Supplier MilliporeSigma Supplier No. MAB8905
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Measles Blend Antibody, hemagglutinin, clones CV1, CV4 is validated for use in immunofluoresence, immunohistochemistry (paraffin) for the detection of Measles Blend.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Measles Blend
Applications Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone CV1, CV4, CV4
Conjugate Unconjugated
Formulation Liquid ascites fluid containing no preservatives.
Host Species Mouse
Immunogen Hemagglutinin protein obtained from Chicago wild type strain of measles
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 12 months in undiluted aliquots.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.