Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MeCP2 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07013MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-013-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-013-MI Supplier MilliporeSigma Supplier No. 07013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-MeCP2 Antibody (rabbit polyclonal antibody) validated in western blotting to detect MeCP2 also known as MeCP-2 protein, mental retardation X-linked 16, mental retardation X-linked 79, methyl CpG binding protein 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MeCP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide.
Gene Symbols AUTSX3; DKFZp686A24160; MRX16; MRX79; MRXS13; MRXSL; MeCP2; OTTHUMP00000026021; PPMX; RTS; RTT
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 465-478 of mouse MeCP2 (C-PRPNREEPVDSRTP) with an N-terminal cysteine added for conjugation purposes.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_004983
Target Species Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.