Learn More
MED24, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human MED24 peptide using ARM Technology.
Supplier: Abnova Corporation H00009862K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
MED24 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
MED24 | |
MED24 | |
A synthetic peptide of human MED24 | |
RUO | |
9862 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
mediator complex subunit 24 | |
MED24 | |
ARC100, CRSP100, CRSP4, DRIP100, KIAA0130, MGC8748, THRAP4, TRAP100 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.