Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mediator complex subunit 27, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018166 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-166 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-166 Supplier Abnova Corporation Supplier No. H00009442D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MED27 protein.

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. [provided by RefSeq

Sequence: MADVINVSVNLEAFSQAISAIQALRSSVSRVFDCLKDGMRNKETLEGREKAFIAHFQDNLHSVNRDLNELERLSNLVGKPSENHPLHNSGLLSLDPVQDKTPLYSQLLQAYKWSNKLQYHAGLASGLLNQQSLKRSANQMGVSAKRRPKAQPTTLVLPPQYVDDVISRIDRMFPEMSIHLSRPNGTSAMLLVTLGKVLKVIVVMRSLFIDRTIVKGYNENVYTEDGKLDIWSKSNYQVFQKVTDHATTALLHYQLPQMPDVVVRSFMTWLRSYIKLFQAPCQRCGKFLQDGLPPTWRDFRTLEAFHDTCRQ

Specifications

Antigen mediator complex subunit 27
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MED27 protein.
Formulation PBS with no preservative; pH 7.4
Gene MED27
Gene Accession No. NM_004269.2
Gene Alias CRAP34/CRSP34/CRSP8/MGC11274/TRAP37
Gene Symbols MED27
Host Species Rabbit
Immunogen MED27 (NP_004260.2, 1 a.a. ∼ 311 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 9442
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.