Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myocyte enhancer factor 2B, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017840 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-840 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-840 Supplier Abnova Corporation Supplier No. H00004207D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MEF2B protein.

This gene represents numerous read-through transcripts that span geneID:729991 and 100271849. Many read-through transcripts are predicted to be nonsense-mediated decay (NMD) candidates, and are thought to be noncoding. Some transcripts are predicted to be capable of translation reinitation at a downstream AUG, resulting in expression of at least one isoform of myocyte enhancer factor 2B (MEF2B) from this read-through locus. At least one additional MEF2B variant and isoform can be expressed from a downstream promoter, and is annotated on geneID:100271849. [provided by RefSeq

Sequence: MGRKKIQISRILDQRNRQVTFTKRKFGLMKKAYELSVLCDCEIALIIFNSANRLFQYASTDMDRVLLKYTEYSEPHESRTNTDILETLKRRGIGLDGPELEPDEGPEEPGEKFRRLAGEGGDPALPRPRLYPAAPAMPSPDVVYGALPPPGCDPSGLGEALPAQSRPSPFRPAAPKAGPPGLVHPLFSPSHLTSKTPPPLYLPTEGRRSDLPGGLAGPRGGLNTSRSLYSGLQNPCSTATPGPPLGSFPFLPGGPPVGAEAWARRVPQPAAPPRRPPQSASSLSASLRPPGAPATFLRPSPIPCSSPGPWQSLCGLGPPCAGCPWPTAGPGRRSPGGTSPERSPGTARARGDPTSLQASSEKTQQ

Specifications

Antigen myocyte enhancer factor 2B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MEF2B protein.
Formulation PBS with no preservative; pH 7.4
Gene MEF2B
Gene Accession No. NM_005919.1
Gene Alias FLJ32599/FLJ46391/MGC189732/MGC189763/RSRFR2
Gene Symbols MEF2B
Host Species Rabbit
Immunogen MEF2B (NP_005910.1, 1 a.a. ∼ 365 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 4207
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.