Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MEF2BNB, Clone: 5A5, Abnova™

Catalog No. 89300349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-349 Supplier Abnova Corporation Supplier No. H00729991M16
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen MEF2BNB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MEF2BNB.
Formulation PBS with no preservative; pH 7.4
Gene BORCS8
Gene Accession No. BC010931.1
Gene Symbols MEF2BNB
Host Species Mouse
Immunogen MEF2BNB (AAH10931.1, 1 a.a. ∼ 119 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729991
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.